Learn More
Apexbio Technology LLC Nutlin-3, 5mg. Cas: 890090-75-2 MFCD: MFCD07784509. MDM2 antagonist,inhibits MDM2-p53 interaction
SDP
Catalog No. 501904908
Encompass_Preferred
Nutlin-3, a tetra-substituted imidazoline, is a potent and selective small-molecule antagonist of murine double minute 2 (MDM2), which occupies the binding site of p53 in MDM2 and consequently prevent MDM2 binding to p53 leading to the disruption of the autoregulator feedback loop Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4908 Supplier Apexbio Technology LLC Supplier No. A42285
Nutlin-3, a tetra-substituted imidazoline, is a potent and selective small-molecule antagonist of murine double minute 2 (MDM2), which occupies the binding site of p53 in MDM2 and consequently prevent MDM2 binding to p53 leading to the disruption of the autoregulator feedback loop Other sizes are available. Please inqury us for quote.