Learn More
Selleck Chemical LLC Nutlin-3 25mg 890090-75-2
SDP
Catalog No. 507968
Encompass_Preferred
Nutlin-3 is a potent and selective Mdm2 (RING finger-dependent ubiquitin protein ligase for itself and p53) antagonist with IC50 of 90 nM in a cell-free assay; stabilizes p73 in p53-deficient cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-796-8 Supplier Selleck Chemical LLC Supplier No. S106125MG
Nutlin-3 is a potent and selective Mdm2 (RING finger-dependent ubiquitin protein ligase for itself and p53) antagonist with IC50 of 90 nM in a cell-free assay; stabilizes p73 in p53-deficient cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.