Learn More
Abcam (+)-Nutlin-3, p53-MDM2 binding inhibitor, 1MG
Catalog No. 502705745
Encompass_Preferred
MW 581.5 Da, Purity >98%. p53-MDM2 binding inhibitor (IC₅₀ = 13 μM). Less potent enantiomer of (-)-Nutlin-3 (Asc-4428 ). Shows antiproliferative effects (IC₅₀ values are 2.2 and 1.3 μM for human skin and murine fibroblasts respectively). Shows antitumor effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5745 Supplier Abcam Supplier No. AB1444261MG
MW 581.5 Da, Purity >98%. p53-MDM2 binding inhibitor (IC₅₀ = 13 μM). Less potent enantiomer of (-)-Nutlin-3 (Asc-4428 ). Shows antiproliferative effects (IC₅₀ values are 2.2 and 1.3 μM for human skin and murine fibroblasts respectively). Shows antitumor effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement