Learn More
Medchemexpress LLC HY-10029 10mg Medchemexpress, Nutlin-3a CAS:675576-98-4 Purity:>98%
SDP
Catalog No. 502028613
Encompass_Preferred
Medchemexpress, HY-10029 10mg Nutlin-3a CAS:675576-98-4 Nutlin-3a, an active enantiomer of Nutlin-3, is a potent murine double minute (MDM2) inhibitor (IC50=90 nM). Nutlin-3a inhibits MDM2-p53 interactions and stabilizes the p53 protein, and induces cell autophagy and apoptosis. Nutlin-3a has the potential for the study of TP53 wild-type ovarian carcinomas[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8613 Supplier Medchemexpress LLC Supplier No. HY1002910MG
Medchemexpress, HY-10029 10mg Nutlin-3a CAS:675576-98-4 Nutlin-3a, an active enantiomer of Nutlin-3, is a potent murine double minute (MDM2) inhibitor (IC50=90 nM). Nutlin-3a inhibits MDM2-p53 interactions and stabilizes the p53 protein, and induces cell autophagy and apoptosis. Nutlin-3a has the potential for the study of TP53 wild-type ovarian carcinomas[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.