Learn More
Selleck Chemical LLC Nutlin-3a
SDP
Catalog No. 5000000162
Encompass_Preferred
Nutlin-3a ((-)-Nutlin-3) the active enantiomer of Nutlin-3 inhibits the p53/MDM2 interaction with IC50 of 90 nM in a cell-free assay Nutlin-3a induces autophagy and apoptosis in a p53-dependent manner
Catalog No. 50-000-00162 Supplier Selleck Chemical LLC Supplier No. S8059100MG
Nutlin-3a ((-)-Nutlin-3) the active enantiomer of Nutlin-3 inhibits the p53/MDM2 interaction with IC50 of 90 nM in a cell-free assay Nutlin-3a induces autophagy and apoptosis in a p53-dependent manner