Learn More
Selleck Chemical LLC Nutlin-3a 25mg 675576-98-4 (-)-Nutlin-3
SDP
Catalog No. 501365618
Encompass_Preferred
Nutlin-3a ((-)-Nutlin-3), the active enantiomer of Nutlin-3, inhibits the p53/MDM2 interaction with IC50 of 90 nM in a cell-free assay. Nutlin-3a induces autophagy and apoptosis in a p53-dependent manner. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5618 Supplier Selleck Chemical LLC Supplier No. S805925MG
Nutlin-3a ((-)-Nutlin-3), the active enantiomer of Nutlin-3, inhibits the p53/MDM2 interaction with IC50 of 90 nM in a cell-free assay. Nutlin-3a induces autophagy and apoptosis in a p53-dependent manner. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.