Learn More
Medchemexpress LLC Nutlin (3a) | 675576-98-4 | 99.1% | 581.49 | 50 MG
SDP
Catalog No. 5000365940
Encompass_Preferred
Nutlin-3a (Rebemadlin) is a potent and selective MDM2 inhibitor, acting as the active enantiomer of Nutlin-3. It functions by inhibiting MDM2-p53 interactions, thereby stabilizing the p53 protein and inducing both cell autophagy and apoptosis. This compound holds potential for research into TP53 wild-type ovarian carcinomas.
  • Potent murine double minute (MDM2) inhibitor with an IC50 of 90 nM.
  • Inhibits MDM2-p53 interactions.
  • Stabilizes the p53 protein.
  • Induces cell autophagy and apoptosis.
  • Potential for the study of TP53 wild-type ovarian carcinomas.
  • Highly selective MDM2 antagonist and p53 inducer.
Catalog No. 50-003-65940 Supplier Medchemexpress LLC Supplier No. 50MGHY10029
Nutlin-3a (Rebemadlin) is a potent and selective MDM2 inhibitor, acting as the active enantiomer of Nutlin-3. It functions by inhibiting MDM2-p53 interactions, thereby stabilizing the p53 protein and inducing both cell autophagy and apoptosis. This compound holds potential for research into TP53 wild-type ovarian carcinomas.
  • Potent murine double minute (MDM2) inhibitor with an IC50 of 90 nM.
  • Inhibits MDM2-p53 interactions.
  • Stabilizes the p53 protein.
  • Induces cell autophagy and apoptosis.
  • Potential for the study of TP53 wild-type ovarian carcinomas.
  • Highly selective MDM2 antagonist and p53 inducer.