Learn More
Medchemexpress LLC Nutlin-3a | 675576-98-4 | MFCD14636430 | 99.1% | 581.49 | C30H30Cl2N4O4 | 25 MG
SDP
Catalog No. 5000214628
Encompass_Preferred
Nutlin-3a is a small-molecule MDM2 antagonist used in research to activate wild-type p53, stabilize p53 protein, and induce apoptosis and autophagy. It is intended for laboratory research use and is characterized for cell-based and biochemical assays.
  • Inhibits MDM2-p53 interaction, leading to p53 activation.
  • Induces apoptosis and autophagy in p53 wild-type cells.
  • High reported purity (≈99.1%) suitable for research applications.
  • Molecular formula C30H30Cl2N4O4 and molecular weight ~581.5 g/mol.
  • Available in multiple solid and solution sizes for experimental flexibility.
  • For research use only; not for human or animal therapeutic use.
Catalog No. 50-002-14628 Supplier Medchemexpress LLC Supplier No. HY1002925MG
Nutlin-3a is a small-molecule MDM2 antagonist used in research to activate wild-type p53, stabilize p53 protein, and induce apoptosis and autophagy. It is intended for laboratory research use and is characterized for cell-based and biochemical assays.
  • Inhibits MDM2-p53 interaction, leading to p53 activation.
  • Induces apoptosis and autophagy in p53 wild-type cells.
  • High reported purity (≈99.1%) suitable for research applications.
  • Molecular formula C30H30Cl2N4O4 and molecular weight ~581.5 g/mol.
  • Available in multiple solid and solution sizes for experimental flexibility.
  • For research use only; not for human or animal therapeutic use.