Learn More
Medchemexpress LLC Nutlin-3a | 675576-98-4 | MFCD14636430 | 99.1% | 581.49 g/mol | C30H30Cl2N4O4 | 1 MG
SDP
Catalog No. 5000213987
Encompass_Preferred
Nutlin-3a is a research-grade small-molecule MDM2 antagonist that stabilizes and activates p53, promoting apoptosis and autophagy in preclinical studies. Supplied as a purified solid for laboratory use, it is intended for cellular and biochemical investigation of p53-dependent pathways.
  • Potent MDM2 inhibitor that activates p53 signaling.
  • High purity suitable for research applications.
  • Characterized molecular weight and formula for analytical work.
  • Available in small quantities for experimental testing.
  • Documented in literature for studies of apoptosis and autophagy.
Catalog No. 50-002-13987 Supplier Medchemexpress LLC Supplier No. HY100291MG
Nutlin-3a is a research-grade small-molecule MDM2 antagonist that stabilizes and activates p53, promoting apoptosis and autophagy in preclinical studies. Supplied as a purified solid for laboratory use, it is intended for cellular and biochemical investigation of p53-dependent pathways.
  • Potent MDM2 inhibitor that activates p53 signaling.
  • High purity suitable for research applications.
  • Characterized molecular weight and formula for analytical work.
  • Available in small quantities for experimental testing.
  • Documented in literature for studies of apoptosis and autophagy.