Learn More
Apexbio Technology LLC Nutlin-3a chiral 675576-98-4 50mg
SDP
Catalog No. 5000062835
Encompass_Preferred
Nutlin-3a chiral (CAS 675576-98-4) is a small-molecule antagonist targeting mouse double minute 2 (MDM2) which functions as a negative regulator of the tumor suppressor TP53 By binding directly to the TP53-interacting pocket of MDM2 Nutlin-3a blocks the degradation of TP53 stabilizing the protein and activating TP53-dependent pathways Studies in solid tumors and lymphomas indicate that Nutlin-3a induces cell cycle arrest growth inhibition and apoptosis Research has demonstrated antitumor activity in vitro and in vivo highlighting its utility in studying TP53-related cell signaling pathways and its potential in cancer research applications
Catalog No. 50-000-62835 Supplier Apexbio Technology LLC Supplier No. A367150
Nutlin-3a chiral (CAS 675576-98-4) is a small-molecule antagonist targeting mouse double minute 2 (MDM2) which functions as a negative regulator of the tumor suppressor TP53 By binding directly to the TP53-interacting pocket of MDM2 Nutlin-3a blocks the degradation of TP53 stabilizing the protein and activating TP53-dependent pathways Studies in solid tumors and lymphomas indicate that Nutlin-3a induces cell cycle arrest growth inhibition and apoptosis Research has demonstrated antitumor activity in vitro and in vivo highlighting its utility in studying TP53-related cell signaling pathways and its potential in cancer research applications