Learn More
Apexbio Technology LLC Nutlin-3b 675576-97-3 5mg
SDP
Catalog No. 502240348
Encompass_Preferred
Nutlin-3b (CAS 675576-97-3) is the inactive enantiomer of the small molecule Nutlin-3 a known MDM2 inhibitor Unlike its active counterpart Nutlin-3a Nutlin-3b exhibits minimal affinity for MDM2 approximately 200-fold lower and lacks the capacity to stabilize wild-type TP53 or influence cell cycle progression proliferation or gene expression associated with TP53 signaling Nutlin-3b does not induce MDM2 p53 or p21 expression nor does it affect colony formation in cancer cell lines such as H460 Due to its negligible biological activity Nutlin-3b serves as a negative control in studies investigating p53 MDM2 interactions and the specific effects of active MDM2 antagonists in cellular and molecular oncology research
This item is not returnable. View return policy
Catalog No. 50-224-0348 Supplier Apexbio Technology LLC Supplier No. A20055
Nutlin-3b (CAS 675576-97-3) is the inactive enantiomer of the small molecule Nutlin-3 a known MDM2 inhibitor Unlike its active counterpart Nutlin-3a Nutlin-3b exhibits minimal affinity for MDM2 approximately 200-fold lower and lacks the capacity to stabilize wild-type TP53 or influence cell cycle progression proliferation or gene expression associated with TP53 signaling Nutlin-3b does not induce MDM2 p53 or p21 expression nor does it affect colony formation in cancer cell lines such as H460 Due to its negligible biological activity Nutlin-3b serves as a negative control in studies investigating p53 MDM2 interactions and the specific effects of active MDM2 antagonists in cellular and molecular oncology research