Learn More
Medchemexpress LLC Nutlin-C1-amido-PEG4-C2-N3 | 99.2% | 883.8 | C42H52Cl2N8O9 | 5 MG
SDP
Catalog No. 5000264913
Encompass_Preferred
Nutlin-C1-amido-PEG4-C2-N3 is a Nutlin-derived ligand-linker conjugate featuring an amido-linked PEG4 spacer and an azide functional group. Supplied as a 5 mg research reagent, it is intended for use in conjugation chemistry and PROTAC design. The product has molecular formula C42H52Cl2N8O9, molecular weight 883.83 g/mol, and HPLC purity 99.19% according to the certificate of analysis.
  • contains an azide functional group compatible with CuAAC click chemistry.
  • peg4 spacer provides hydrophilic linker length for conjugation.
  • high purity (99.19% by HPLC) suitable for research applications.
  • provided in a small, pre-weighed quantity for building-block synthesis.
  • certificate of analysis available for batch-specific verification.
Catalog No. 50-002-64913 Supplier Medchemexpress LLC Supplier No. HY1288325MG
Nutlin-C1-amido-PEG4-C2-N3 is a Nutlin-derived ligand-linker conjugate featuring an amido-linked PEG4 spacer and an azide functional group. Supplied as a 5 mg research reagent, it is intended for use in conjugation chemistry and PROTAC design. The product has molecular formula C42H52Cl2N8O9, molecular weight 883.83 g/mol, and HPLC purity 99.19% according to the certificate of analysis.
  • contains an azide functional group compatible with CuAAC click chemistry.
  • peg4 spacer provides hydrophilic linker length for conjugation.
  • high purity (99.19% by HPLC) suitable for research applications.
  • provided in a small, pre-weighed quantity for building-block synthesis.
  • certificate of analysis available for batch-specific verification.