Learn More
Medchemexpress LLC Nutlin carboxylic acid | 2249750-27-2 | 98.6% | 100 MG
SDP
Catalog No. 5000338912
Encompass_Preferred
Nutlin carboxylic acid is a Nutlin 3-based MDM2 ligand designed to form PROTACs (Proteolysis-Targeting Chimeras). These PROTACs operate by harnessing the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. It has a molecular weight of 639.53 and a chemical formula of C32H32Cl2N4O6, appearing as a white to yellow solid.
  • Nutlin 3-based MDM2 ligand
  • Forms PROTACs via a linker
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-38912 Supplier Medchemexpress LLC Supplier No. HY128837100MG
Nutlin carboxylic acid is a Nutlin 3-based MDM2 ligand designed to form PROTACs (Proteolysis-Targeting Chimeras). These PROTACs operate by harnessing the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins. It has a molecular weight of 639.53 and a chemical formula of C32H32Cl2N4O6, appearing as a white to yellow solid.
  • Nutlin 3-based MDM2 ligand
  • Forms PROTACs via a linker
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only