Learn More
Medchemexpress LLC HY-128837 5mg Medchemexpress, Nutlin carboxylic acid CAS:2249750-27-2 Purity:>98%
SDP
Catalog No. 501969775
Encompass_Preferred
Medchemexpress, HY-128837 5mg Nutlin carboxylic acid CAS:2249750-27-2 Nutlin carboxylic acid (MDM2 ligand 1) is the Nutlin 3-based MDM2 ligand. Nutlin carboxylic acid (MDM2 ligand 1) can be connected to the ligand for protein by a linker to form PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9775 Supplier Medchemexpress LLC Supplier No. HY1288375MG
Medchemexpress, HY-128837 5mg Nutlin carboxylic acid CAS:2249750-27-2 Nutlin carboxylic acid (MDM2 ligand 1) is the Nutlin 3-based MDM2 ligand. Nutlin carboxylic acid (MDM2 ligand 1) can be connected to the ligand for protein by a linker to form PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.