Learn More
Cayman Chemical Nutln-3 10mg
SDP
Catalog No. 502980127
Encompass_Preferred
An inhibitor of the p53-Mdm2 interaction (IC50 = 0.09 µM); increases p53 levels, induces apoptosis, and reduces cell viability in HCT116, SJSA-1, and RKO cells; reduces tumor volume in SJSA-1, MHM, LNCaP, and 22Rv1 mouse xenograft models at 200 mg/kg
Catalog No. 50-298-0127 Supplier Cayman Chemical Supplier No. 1858510 MG
An inhibitor of the p53-Mdm2 interaction (IC50 = 0.09 µM); increases p53 levels, induces apoptosis, and reduces cell viability in HCT116, SJSA-1, and RKO cells; reduces tumor volume in SJSA-1, MHM, LNCaP, and 22Rv1 mouse xenograft models at 200 mg/kg