Learn More
Bio Serv Bio-Serv Nutra-Gel Bacon, Complete Nutritional Gelled Diet, 1 oz
SDP
Catalog No. NC1993060
Encompass
Bio-Serv Nutra-Gel Bacon offers a highly palatable, nutritionally complete gel diet for rodents, designed for various research applications. Packaged in convenient, ready-to-serve 1 oz cups, it provides both food and water and is ideal for animals with mobility impairments or those recovering from health issues. Its unique formulation and sterilization ensure quality, making it a standout option for researchers.
Catalog No. NC1993060 Supplier Bio Serv Supplier No. NGB1
Bio-Serv Nutra-Gel Bacon offers a highly palatable, nutritionally complete gel diet for rodents, designed for various research applications. Packaged in convenient, ready-to-serve 1 oz cups, it provides both food and water and is ideal for animals with mobility impairments or those recovering from health issues. Its unique formulation and sterilization ensure quality, making it a standout option for researchers.