Learn More
Sigma Aldrich Fine Chemicals Biosciences NutridomaTM CS
Catalog No. 502905915
Encompass
Nutridoma-CS is a serum replacement SOLUTIONon which helps in cell proliferation when added to a suspension. Its major component is -D xylopyranose which has ring shaped sugar allowing the cells to grow in a suspension even without serum.
Catalog No. 50-290-5915 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11363743001
Nutridoma-CS is a serum replacement SOLUTIONon which helps in cell proliferation when added to a suspension. Its major component is -D xylopyranose which has ring shaped sugar allowing the cells to grow in a suspension even without serum.