Learn More
NEOGEN Nutrient Agar 1.5%, 10 kg
SDP
Catalog No. 502015776
Encompass_Preferred
Nutrient Agar 1.5% is used for the cultivation of a wide variety of microorganisms in a laboratory setting. Nutrient Agar 1.5% is not intended for use in the diagnosis of disease or other conditions in humans. In the early 1900’s, the American Public Health Association (APHA) suggested the formula of Nutrient Agar as a standard culture medium used in water testing. Nutrient Agar is specified in many standard method procedures. Nutrient Agar 1.5% is a modification of Nutrient Agar. Nutrient Agar 1.5% is a general purpose medium, with a slightly alkaline pH. This medium contains 0.8% sodium chloride and can be used as a base for enrichment with blood, ascitic fluid, or other supplements for cultivating fastidious microorganisms.For more information visit https://www.neogen.com/solutions/Microbiology/nutrient-agar-1-5-percent
Catalog No. 50-201-5776 Supplier NEOGEN Supplier No. 700003554
Nutrient Agar 1.5% is used for the cultivation of a wide variety of microorganisms in a laboratory setting. Nutrient Agar 1.5% is not intended for use in the diagnosis of disease or other conditions in humans. In the early 1900’s, the American Public Health Association (APHA) suggested the formula of Nutrient Agar as a standard culture medium used in water testing. Nutrient Agar is specified in many standard method procedures. Nutrient Agar 1.5% is a modification of Nutrient Agar. Nutrient Agar 1.5% is a general purpose medium, with a slightly alkaline pH. This medium contains 0.8% sodium chloride and can be used as a base for enrichment with blood, ascitic fluid, or other supplements for cultivating fastidious microorganisms.For more information visit https://www.neogen.com/solutions/Microbiology/nutrient-agar-1-5-percent