Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Nutrient Agar
SCI_ED

Catalog No. S29562 Shop All Aldon Products
Change view
Click to view available options
Quantity:
125 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S29562 125 mL
S29561 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S29562 Supplier Innovating Science™ Supplier No. MD0101CS/24
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality microbiological media at an economical price

  • Innovating Science™ Sterile Prepared Agar and Broth will bring quality back to the lab market
  • Case of 24
  • Specifications

    Certifications/Compliance Sterile
    Concentration 1
    Content And Storage Room Temperature
    Form Gel
    Format Bottle
    Manufacturing Origin United States
    Media Type Nutrient Agar
    Product Type Prepared Microbiology Media
    Sterility Sterile
    Target Organism Class Bacteria
    Test Type Microbial Detection
    For Use With (Application) Microbes
    For Use With (Equipment) Microbes, Educational
    Packaging Type Glass
    Quantity 125 mL
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.