Learn More
Research Products International Corp Nutrient Agar, 2 KG
SDP
Catalog No. 501977334
Encompass_Preferred
For cultivating a variety of non-fastidious organisms.
Dissolve 23g in one liter distilled water. Heat solution to boiling with continuous stirring for one minute. Sterilize by autoclaving at 121°C for 15 minutes. Mix medium with gentle agitation to avoid bubbles prior to pouring plates. Store at room temperature.
Ingredients:
Beef Extract: 3 g/l
Pancreatic Digest of Gelatin: 5 g/l
Agar: 15 g/l
Catalog No. 50-197-7334 Supplier Research Products International Corp Supplier No. N141002000.0
For cultivating a variety of non-fastidious organisms.
Dissolve 23g in one liter distilled water. Heat solution to boiling with continuous stirring for one minute. Sterilize by autoclaving at 121°C for 15 minutes. Mix medium with gentle agitation to avoid bubbles prior to pouring plates. Store at room temperature.
Ingredients:
Beef Extract: 3 g/l
Pancreatic Digest of Gelatin: 5 g/l
Agar: 15 g/l