Learn More
Alkali Scientific Nutrient Agar, 20 mL, 100 × 15 mm Plate, 10/Pk
SDP
Catalog No. 5000663032
Encompass_Preferred
Nutrient Agar, 20 mL, 100 × 15 mm Plate, 10/Pk. Nutrient Agar is a versatile medium that supports the growth of a wide range of non-fibrous bacteria. It is used in microbiological labs for general bacterial culturing, providing the essential nutrients required for optimal growth. The pre-prepared plate format ensures ease of use and consistency, making it a convenient tool for routine microbiological work.
Catalog No. 50-006-63032 Supplier Alkali Scientific Supplier No. NTRAP00
Nutrient Agar, 20 mL, 100 × 15 mm Plate, 10/Pk. Nutrient Agar is a versatile medium that supports the growth of a wide range of non-fibrous bacteria. It is used in microbiological labs for general bacterial culturing, providing the essential nutrients required for optimal growth. The pre-prepared plate format ensures ease of use and consistency, making it a convenient tool for routine microbiological work.