Learn More
Bioworld Nutrient Agar, 500 g
Supplier:  Bioworld 306200551
Encompass_Preferred
Nutrient AgarRecommended for the cultivation of non-fastidious microorganisms according to the methods described by the American Public Health AssociationpH: 6.8Prepared Appearance: light amber agarContains (g/L):Beef Extract - 3.0g Pancreatic Digest of Gelatin - 5.0g Agar - 15.0g
Catalog No. 50-254-339