Learn More
NEOGEN Nutrient Agar, 500 g
SDP
Catalog No. 502015249
Encompass_Preferred
Nutrient Agar is used for the cultivation of a wide variety of microorganisms and is not intended for use in the diagnosis of disease or other conditions in humans. A general purpose medium for the cultivation of organisms that are not demanding in their nutritional requirements e.g. organisms that can be isolated from air, water, dust etc. Nutrient Agar is suitable for teaching and demonstration purposes, it is isotonic and can be enriched with biological fluids such as sterile blood and egg yolk.  Formula:Peptone - 5.0 g/LMeat Extract - 3.0 g/LSodium Chloride - 8.0 g/LAgar - 12.0 g/L For more information visit https://www.neogen.com/solutions/Microbiology/nutrient-agar
Catalog No. 50-201-5249 Supplier NEOGEN Supplier No. 700004402
Nutrient Agar is used for the cultivation of a wide variety of microorganisms and is not intended for use in the diagnosis of disease or other conditions in humans. A general purpose medium for the cultivation of organisms that are not demanding in their nutritional requirements e.g. organisms that can be isolated from air, water, dust etc. Nutrient Agar is suitable for teaching and demonstration purposes, it is isotonic and can be enriched with biological fluids such as sterile blood and egg yolk.  Formula:Peptone - 5.0 g/LMeat Extract - 3.0 g/LSodium Chloride - 8.0 g/LAgar - 12.0 g/L For more information visit https://www.neogen.com/solutions/Microbiology/nutrient-agar