Learn More
MOLTOX Molecular Toxicology Inc Nutrient Agar, 20 mL/plate, 10 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948825
Encompass_Preferred
Nutrient Agar is a general purpose medium for the cultivation and maintenance of nonfastidious organisms. It can be used for the cultivation and enumeration of organisms in water, sewage, feces and other materials. Nutrient agars prepared with Oxoid Nutrient Broth No. 2, specified by Maron & Ames, Mutat. Res. 113: 173, 1983, are used for general growth and CFU determinations of Ames S. typhimurium and E. coli WP2 strains. We provide specifically formulated Nutrient agars and broths for Ames assay applications as well as for general use. MOLTOX does not provide microorganisms for testing. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.
Catalog No. 50-948-825 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S031
Nutrient Agar is a general purpose medium for the cultivation and maintenance of nonfastidious organisms. It can be used for the cultivation and enumeration of organisms in water, sewage, feces and other materials. Nutrient agars prepared with Oxoid Nutrient Broth No. 2, specified by Maron & Ames, Mutat. Res. 113: 173, 1983, are used for general growth and CFU determinations of Ames S. typhimurium and E. coli WP2 strains. We provide specifically formulated Nutrient agars and broths for Ames assay applications as well as for general use. MOLTOX does not provide microorganisms for testing. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.