Learn More
MOLTOX Molecular Toxicology Inc Nutrient Agar, 7mL/tube, 10 tubes/box, Store in Refrigerator
SDP
Catalog No. 50948721
Encompass_Preferred
Nutrient Agar is a general purpose medium for the cultivation and maintenance of nonfastidious organisms. It can be used for the cultivation and enumeration of organisms in water, sewage, feces and other materials. Nutrient agars prepared with Oxoid Nutrient Broth No. 2, specified by Maron & Ames, Mutat. Res. 113: 173, 1983, are used for general growth and CFU determinations of Ames S. typhimurium and E. coli WP2 strains. We provide specifically formulated Nutrient agars and broths for Ames assay applications as well as for general use. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.
Catalog No. 50-948-721 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 22132.007H
Nutrient Agar is a general purpose medium for the cultivation and maintenance of nonfastidious organisms. It can be used for the cultivation and enumeration of organisms in water, sewage, feces and other materials. Nutrient agars prepared with Oxoid Nutrient Broth No. 2, specified by Maron & Ames, Mutat. Res. 113: 173, 1983, are used for general growth and CFU determinations of Ames S. typhimurium and E. coli WP2 strains. We provide specifically formulated Nutrient agars and broths for Ames assay applications as well as for general use. Some media are temperature sensitive and may have specific temperature and storage requirements for shipping/delivery.