Learn More
Alkali Scientific Nutrient Agar Slant 10mL Tube, 10/Pk
SDP
Catalog No. 5000663030
Encompass_Preferred
Nutrient Agar Slants (10 mL) are general purpose culture media used for cultivating and maintaining a wide variety of non fastidious microorganisms. The larger 10 mL fill provides an expanded slanted surface and deeper agar butt compared with standard slants. This increased volume helps retain moisture for longer periods, making the tubes suitable for extended storage of stock cultures. The medium contains beef extract and peptone that supply essential nutrients including amino acids, vitamins, and organic nitrogen compounds. The slanted surface allows aerobic growth while the deeper portion of the medium helps maintain hydration. These slants are widely used in microbiology laboratories for culture maintenance, biomass generation, and preparation of sub cultures.
Catalog No. 50-006-63030 Supplier Alkali Scientific Supplier No. NNUT01
Nutrient Agar Slants (10 mL) are general purpose culture media used for cultivating and maintaining a wide variety of non fastidious microorganisms. The larger 10 mL fill provides an expanded slanted surface and deeper agar butt compared with standard slants. This increased volume helps retain moisture for longer periods, making the tubes suitable for extended storage of stock cultures. The medium contains beef extract and peptone that supply essential nutrients including amino acids, vitamins, and organic nitrogen compounds. The slanted surface allows aerobic growth while the deeper portion of the medium helps maintain hydration. These slants are widely used in microbiology laboratories for culture maintenance, biomass generation, and preparation of sub cultures.