Learn More
Hach Company BOD Nutrient Buffer Pillows, 3 mL concentrate to make 3L, 50/pk
Catalog No. 501995136 Shop All Hach Company Products
Encompass
For preparation of BOD5 dilution water. Each pillow combines the APHA-specified buffer and nutrients to prepare 3 liters of dilution water. Pack of 50 pillows
Catalog No. 50-199-5136 Supplier Hach Company Supplier No. 1486166
For preparation of BOD5 dilution water. Each pillow combines the APHA-specified buffer and nutrients to prepare 3 liters of dilution water. Pack of 50 pillows