Learn More
Hach Company BOD Nutrient Buffer Pillows, 6 mL concentrate to make 6 L, 50/pk
Catalog No. 501995137 Shop All Hach Company Products
Encompass
For preparation of BOD5 dilution water. Each pillow combines the APHA-specified buffer andnutrients to prepare 6 liters of dilution water. Pack of 50 pillows.
Catalog No. 50-199-5137 Supplier Hach Company Supplier No. 1486266
For preparation of BOD5 dilution water. Each pillow combines the APHA-specified buffer andnutrients to prepare 6 liters of dilution water. Pack of 50 pillows.