Learn More
Hach Company BOD Nutrient Buffer Pillows, 19 mL of concentrate to make 19 L, 25/pk
Catalog No. 501995138 Shop All Hach Company Products
Encompass
For preparation of BOD5 dilution water. Each pillow combines the APHA-specified buffer andnutrients to prepare 19 liters of dilution water. Pack of 25 pillows.
Catalog No. 50-199-5138 Supplier Hach Company Supplier No. 1486398
For preparation of BOD5 dilution water. Each pillow combines the APHA-specified buffer andnutrients to prepare 19 liters of dilution water. Pack of 25 pillows.