Learn More
Alkali Scientific Nutrient Broth 200mL Bottle, 1/Pk
SDP
Catalog No. 5000663065
Encompass_Preferred
Nutrient Broth is a classic liquid culture medium used for the growth of non fastidious microorganisms. The 200 mL bottle format provides a convenient sterile volume suitable for preparing larger bacterial cultures or dispensing into smaller laboratory vessels. The formulation contains beef extract and peptone, which supply essential nutrients including amino acids, vitamins, carbohydrates, and nitrogen compounds that support microbial growth. Because the medium is non selective, it can cultivate a wide range of organisms including Enterobacteriaceae, Bacillus, and Staphylococcus species. Nutrient broth is commonly used for biomass production, enrichment of microbial samples, and preparation of bacterial suspensions for laboratory testing.
Catalog No. 50-006-63065 Supplier Alkali Scientific Supplier No. NNUB00
Nutrient Broth is a classic liquid culture medium used for the growth of non fastidious microorganisms. The 200 mL bottle format provides a convenient sterile volume suitable for preparing larger bacterial cultures or dispensing into smaller laboratory vessels. The formulation contains beef extract and peptone, which supply essential nutrients including amino acids, vitamins, carbohydrates, and nitrogen compounds that support microbial growth. Because the medium is non selective, it can cultivate a wide range of organisms including Enterobacteriaceae, Bacillus, and Staphylococcus species. Nutrient broth is commonly used for biomass production, enrichment of microbial samples, and preparation of bacterial suspensions for laboratory testing.