Learn More
Bioworld Nutrient Broth, 5 kg
Catalog No. 50254342
Encompass_Preferred
Nutrient BrothRecommended for the cultivation of non-fastidious microorganisms according to the methods described by the American Public Health AssociationpH: 6.8Working Concentration: 8 g/LDehydrated Appearance: medium tan, homogeneous powderPrepared Appearance: light to medium amber brothSoluble in waterComposition (g/L):Beef Extrac - 3.0g Pancreatic Digest of Gelatin- 5.0g
Catalog No. 50-254-342 Supplier Bioworld Supplier No. 306200563
Nutrient BrothRecommended for the cultivation of non-fastidious microorganisms according to the methods described by the American Public Health AssociationpH: 6.8Working Concentration: 8 g/LDehydrated Appearance: medium tan, homogeneous powderPrepared Appearance: light to medium amber brothSoluble in waterComposition (g/L):Beef Extrac - 3.0g Pancreatic Digest of Gelatin- 5.0g