Learn More
Research Products International Corp Nutrient Broth, 5 Kilograms
SDP
Catalog No. 50488847
Encompass_Preferred
For cultivating a variety of non-fastidious organisms.
Dissolve 8 grams in one liter distilled water. Heat solution to boiling with continuous stirring for one minute. Sterilize by autoclaving at 121°C for 15 minutes. Store at room temperature.
Beef Extract: 3 g/l
Pancreatic Digest of Gelatin: 5 g/l
Catalog No. 50-488-847 Supplier Research Products International Corp Supplier No. N151005000.0
For cultivating a variety of non-fastidious organisms.
Dissolve 8 grams in one liter distilled water. Heat solution to boiling with continuous stirring for one minute. Sterilize by autoclaving at 121°C for 15 minutes. Store at room temperature.
Beef Extract: 3 g/l
Pancreatic Digest of Gelatin: 5 g/l