Learn More
Research Products International Corp Nutrient Broth, 500 Grams
SDP
Catalog No. 50488846
Encompass_Preferred
For cultivating a variety of non-fastidious organisms.
Dissolve 8 grams in one liter distilled water. Heat solution to boiling with continuous stirring for one minute. Sterilize by autoclaving at 121°C for 15 minutes. Store at room temperature.
Beef Extract: 3 g/l
Pancreatic Digest of Gelatin: 5 g/l
Catalog No. 50-488-846 Supplier Research Products International Corp Supplier No. N15100500.0
For cultivating a variety of non-fastidious organisms.
Dissolve 8 grams in one liter distilled water. Heat solution to boiling with continuous stirring for one minute. Sterilize by autoclaving at 121°C for 15 minutes. Store at room temperature.
Beef Extract: 3 g/l
Pancreatic Digest of Gelatin: 5 g/l