Learn More
NEOGEN Nutrient Broth, 500 g
SDP
Supplier:  NEOGEN 700003276
Encompass_Preferred
Nutrient Broth is used for the cultivation of a wide variety of microorganisms. Nutrient Broth is not intended for use in the diagnosis of disease or other conditions in humans.In the early 1900’s, the American Public Health Association (APHA) suggested the formula of Nutrient Agar as a standard culture medium used in water testing. Nutrient Broth is the same formulation as Nutrient Agar, only Agar has been omitted.Nutrient Broth is used as a pre-enrichment medium when testing certain foods and dairy products for Salmonella spp. In dried or processed foods, salmonellae may be sub-lethally injured and in low numbers. The presence of other bacteria and food sample components may hinder growth and recovery of Salmonella spp.For more information visit https://www.neogen.com/solutions/Microbiology/nutrient-broth
Catalog No. 50-201-5468