Learn More
NEOGEN Nutrient Broth No. 2, 500 g
SDP
Catalog No. 502015687
Encompass_Preferred
Nutrient Both No. 2 is used for the cultivation of fastidious bacteria, and forms the base for Preston Broth according to ISO 10272-1:2017 for the cultivation of Campylobacter spp. This media is not intended for use in the diagnosis of disease or other conditions in humans. This broth can also be used as the suspending medium for cooked meat granules for the cultivation of anaerobic organisms.For more information visit https://www.neogen.com/solutions/Microbiology/nutrient-broth-number-2
Catalog No. 50-201-5687 Supplier NEOGEN Supplier No. 700004597
Nutrient Both No. 2 is used for the cultivation of fastidious bacteria, and forms the base for Preston Broth according to ISO 10272-1:2017 for the cultivation of Campylobacter spp. This media is not intended for use in the diagnosis of disease or other conditions in humans. This broth can also be used as the suspending medium for cooked meat granules for the cultivation of anaerobic organisms.For more information visit https://www.neogen.com/solutions/Microbiology/nutrient-broth-number-2