Learn More
MOLTOX Molecular Toxicology Inc Nutrient Broth Oxoid # 2. 50ml/bottle. Store at Room Temperature.
SDP
Supplier:  MOLTOX Molecular Toxicology Inc 26505.05
Encompass
Nutrient Broth is used for the cultivation of various nonfastidious microorganisms. The formulations which include Oxoid Nutrient Broth No. 2 are primarily used in the Ames assay as specified by Maron & Ames, Mutat. Res. 113: 173, 1983 for culture of S. typhimurium. Also used for culture of E. coli WP2. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. NC1399185