Learn More
Description
Reduce media preparation time and cultivate fewer fastidious microorganisms using convenient, pre-filled, ready-to-use Thermo Scientific™ Nutrient Broth.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of Selenite Broth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Clear liquid, light amber |
| Content And Storage | 2°C to 8°C |
| Format | Tube |
| Manufacturing Origin | United States |
| Product Type | Broth Tube |
| Fill Volume | 5 mL |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.