Learn More
Alkali Scientific Nutrient Gelatin Deep 8mL Tube, 10/Pk
SDP
Catalog No. 5000663063
Encompass_Preferred
Nutrient Gelatin is a differential medium used to determine whether microorganisms produce gelatinase, an enzyme capable of hydrolyzing gelatin proteins. Each tube contains approximately 8 mL of medium that solidifies into a deep format and is inoculated by a straight stab through the center. Unlike agar media, gelatin melts at incubation temperatures, so results are interpreted after refrigeration. Following incubation, tubes are cooled to approximately 4°C for 15–30 minutes. If the organism produces gelatinase, the gelatin is hydrolyzed and the medium remains liquid after cooling. If the enzyme is absent, the gelatin solidifies again. This reaction helps identify organisms with proteolytic activity and is commonly used in classical bacterial identification.
Catalog No. 50-006-63063 Supplier Alkali Scientific Supplier No. NGDT00
Nutrient Gelatin is a differential medium used to determine whether microorganisms produce gelatinase, an enzyme capable of hydrolyzing gelatin proteins. Each tube contains approximately 8 mL of medium that solidifies into a deep format and is inoculated by a straight stab through the center. Unlike agar media, gelatin melts at incubation temperatures, so results are interpreted after refrigeration. Following incubation, tubes are cooled to approximately 4°C for 15–30 minutes. If the organism produces gelatinase, the gelatin is hydrolyzed and the medium remains liquid after cooling. If the enzyme is absent, the gelatin solidifies again. This reaction helps identify organisms with proteolytic activity and is commonly used in classical bacterial identification.