Learn More
Sigma Aldrich Fine Chemicals Biosciences Nutrient Mixture F-10 Ham With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture | MFCD00217607 | 500ML
Catalog No. NC1598189
Encompass
Nutrient Mixture F-10 Ham With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture | MFCD00217607 | 500ML
Catalog No. NC1598189 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N6013500ML
Nutrient Mixture F-10 Ham With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture | MFCD00217607 | 500ML