Learn More
Sigma Aldrich Fine Chemicals Biosciences Nutrient Mixture f-12 Ham1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences N352010X1L
Encompass
The nutrient mixture Hams f-12 is developed by Ham and it has higher levels of amino acids vitamins and trace elements. Putrescine and linoleic acid are added to the formulation. Nutrient medium was originally designed for the serum-free growth of Chinese hamster ovary and lung cells. When used with whole or dialyzed serum or in combination with hormones and transferrin it is widely used to grow mammalian and hybridoma cells.
Catalog No. 50-290-1885