Learn More
Reprocell NUTRISTEM XF/FF CULTURE MEDIUM
Catalog No. NC0345861
Encompass
NutriStem XF/FF Culture Medium for Human iPS and ES Cells, 500ml. NutriStem XF/FF Culture Medium is a fully defined xeno-free, low growth factor human embryonic stem (ES) and induced pluripotent stem (iPS) cell feeder-free culture medium that enables maintenance and expansion of pluripotent stem cells. NutriStem Medium can be used to culture pluripotent stem cells for at least 20 passages while retaining pluripotency marker expression, robust proliferation with a normal karyotype, and the ability to differentiate into cells of all three germ layers in vitro and in vivo. NutriStem Medium offers the ability to culture cells in a completely xeno-free medium without the need for high levels of basic FGF and other stimulatory growth factors and cytokines. In addition, the superior cell attachment and proliferation observed with NutriStem Medium aid high-throughput screening applications.
Catalog No. NC0345861 Supplier Reprocell Supplier No. 010005
NutriStem XF/FF Culture Medium for Human iPS and ES Cells, 500ml. NutriStem XF/FF Culture Medium is a fully defined xeno-free, low growth factor human embryonic stem (ES) and induced pluripotent stem (iPS) cell feeder-free culture medium that enables maintenance and expansion of pluripotent stem cells. NutriStem Medium can be used to culture pluripotent stem cells for at least 20 passages while retaining pluripotency marker expression, robust proliferation with a normal karyotype, and the ability to differentiate into cells of all three germ layers in vitro and in vivo. NutriStem Medium offers the ability to culture cells in a completely xeno-free medium without the need for high levels of basic FGF and other stimulatory growth factors and cytokines. In addition, the superior cell attachment and proliferation observed with NutriStem Medium aid high-throughput screening applications.