Learn More
Cayman Chemical Neutrophil Elastase Inhibitor, Purity: ≥95%, CAS: 1448314-31-5, 10 mg
SDP
Catalog No. 502971107
Encompass_Preferred
An N-benzoylindazole derivative that selectively targets the binding domain of neutrophil elastase (IC50 = 7 nM)
Catalog No. 50-297-1107 Supplier Cayman Chemical Supplier No. 1492210 MG
An N-benzoylindazole derivative that selectively targets the binding domain of neutrophil elastase (IC50 = 7 nM)