Learn More
Cayman Chemical nutrophIl Elasts InhIbItr 25mg
SDP
Catalog No. 502969480
Encompass_Preferred
An N-benzoylindazole derivative that selectively targets the binding domain of neutrophil elastase (IC50 = 7 nM)
Catalog No. 50-296-9480 Supplier Cayman Chemical Supplier No. 1492225 MG
Add to Cart
Add to Cart
An N-benzoylindazole derivative that selectively targets the binding domain of neutrophil elastase (IC50 = 7 nM)