Learn More
Cayman Chemical nutrophIlmouse IsolatIon 1ea
SDP
Catalog No. 502894651
Encompass_Preferred
An inhibitor of the ERCC-1-XPF protein-protein interaction (IC50 1.86 UM) an inhibitor of D-Ala-D-Ala ligase (Ki 218 UM for the E. coli enzyme) active against E. coli and S. aureus (MICs 64 and 32 UG/ml respectively) potentiates cisplatin-induced cytotoxicity in A549 and HCT116 cancer cells at 100 UM
Catalog No. 50-289-4651 Supplier Cayman Chemical Supplier No. 6010701 EA
An inhibitor of the ERCC-1-XPF protein-protein interaction (IC50 1.86 UM) an inhibitor of D-Ala-D-Ala ligase (Ki 218 UM for the E. coli enzyme) active against E. coli and S. aureus (MICs 64 and 32 UG/ml respectively) potentiates cisplatin-induced cytotoxicity in A549 and HCT116 cancer cells at 100 UM