Learn More
Chemglass Life Sciences Chemglass Nut, Flangeless, 1/16 in, Yellow, Tefzel - for flangeless ferrule and FEP tubing
SDP
Catalog No. NC1756617
Encompass_Preferred
Flangeless nut designed for 1/16 in tubing, molded from Tefzel for broad chemical resistance. Used with a flangeless ferrule to create a low-profile, leak-resistant connection to 1/4 in-28 fittings. Sold individually.
  • Made from Tefzel for chemical resistance.
  • Fits 1/16 in tubing.
  • Yellow color for size identification.
  • 1/4 in-28 thread for standard fittings.
  • Simplifies installation when used with a flangeless ferrule.
  • Sold individually (each).
Catalog No. NC1756617 Supplier Chemglass Life Sciences Supplier No. CG1164N01
Flangeless nut designed for 1/16 in tubing, molded from Tefzel for broad chemical resistance. Used with a flangeless ferrule to create a low-profile, leak-resistant connection to 1/4 in-28 fittings. Sold individually.
  • Made from Tefzel for chemical resistance.
  • Fits 1/16 in tubing.
  • Yellow color for size identification.
  • 1/4 in-28 thread for standard fittings.
  • Simplifies installation when used with a flangeless ferrule.
  • Sold individually (each).