Learn More
Microsolv Technology Corporation Nuts Lp Delrin Ff Green 1/8
Catalog No. 5001451484
Encompass_Preferred
Tubing Connector Fittings, Low Pressure, 1/8th, Delrin, Green with 1/4-28 Screw Threads, for use with Flange Free, polymeric tubing. MicroSolv Brand. 10/PK.
Catalog No. 50-014-51484 Supplier Microsolv Technology Corporation Supplier No. 45208G
Tubing Connector Fittings, Low Pressure, 1/8th, Delrin, Green with 1/4-28 Screw Threads, for use with Flange Free, polymeric tubing. MicroSolv Brand. 10/PK.