Learn More
Microsolv Technology Corporation Nuts Lp Delrin Ff Red 1/16
Catalog No. 5001451494
Encompass_Preferred
Tubing Connector Fittings, Low Pressure, 1/16th, Delrin, Red with 1/4-28 Screw Threads, for use with Flange Free, polymeric tubing. MicroSolv Brand. 10/PK.
Catalog No. 50-014-51494 Supplier Microsolv Technology Corporation Supplier No. 45216R
Tubing Connector Fittings, Low Pressure, 1/16th, Delrin, Red with 1/4-28 Screw Threads, for use with Flange Free, polymeric tubing. MicroSolv Brand. 10/PK.