Learn More
Microsolv Technology Corporation Nuts Lp Delrin Ff Red 1/8
Catalog No. 5001451486
Encompass_Preferred
Tubing Connector Fittings, Low Pressure, 1/8th, Delrin, Red with 1/4-28 Screw Threads, for use with Flange Free, polymeric tubing. MicroSolv Brand. 10/PK.
Catalog No. 50-014-51486 Supplier Microsolv Technology Corporation Supplier No. 45208R
Tubing Connector Fittings, Low Pressure, 1/8th, Delrin, Red with 1/4-28 Screw Threads, for use with Flange Free, polymeric tubing. MicroSolv Brand. 10/PK.