Learn More
Microsolv Technology Corporation Nuts Lp Pp Red 1/16Th Male
Catalog No. 5001451468
Encompass_Preferred
Tubing Connector Fittings, Low Pressure, 1/16th, Polypropylene, Red with 1/4-28 Screw Threads, for use with flanged, polymeric tubing. MicroSolv Brand. 10/PK.
Catalog No. 50-014-51468 Supplier Microsolv Technology Corporation Supplier No. 45016R
Tubing Connector Fittings, Low Pressure, 1/16th, Polypropylene, Red with 1/4-28 Screw Threads, for use with flanged, polymeric tubing. MicroSolv Brand. 10/PK.