Learn More
Grainger Milwaukee Tool Magnetic Nut Driver Set, Custom Alloy76™ Steel, Impact Rated, 5/Pk.
Catalog No. 502783047
Encompass_Preferred
These SHOCKWAVE™ Impact Duty™ insert magnetic nut drivers are engineered for heavy-duty fastening tasks. Compatible with bit holders, they are optimized for use with impact drivers and drill drivers, providing reliable performance in a variety of material applications including wood, PVC, aluminum, and sheet metal.
  • Engineered with Custom Alloy76™ steel to extend service life
  • Optimized SHOCK ZONE absorbs peak torque to prevent bit breakage
  • Strong magnetic tip secures screws during application set-up
  • Impact Duty™ magnet prevents loosening over time
  • Improved hex geometry ensures best fit and reduced stripping
  • Chamfered edge design provides maximum screw-head engagement
  • Color-coded bands aid in quick size identification
Catalog No. 50-278-3047 Supplier Grainger Supplier No. 19RK03
These SHOCKWAVE™ Impact Duty™ insert magnetic nut drivers are engineered for heavy-duty fastening tasks. Compatible with bit holders, they are optimized for use with impact drivers and drill drivers, providing reliable performance in a variety of material applications including wood, PVC, aluminum, and sheet metal.
  • Engineered with Custom Alloy76™ steel to extend service life
  • Optimized SHOCK ZONE absorbs peak torque to prevent bit breakage
  • Strong magnetic tip secures screws during application set-up
  • Impact Duty™ magnet prevents loosening over time
  • Improved hex geometry ensures best fit and reduced stripping
  • Chamfered edge design provides maximum screw-head engagement
  • Color-coded bands aid in quick size identification